missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CDK4/Cyclin D1, GST Tag Recombinant Protein
Human CDK4/Cyclin D1, GST Tag Recombinant Protein
€ 624.00 - € 13780.00
Specifications
Accession Number | NP_000066 I NP_4444284 |
---|---|
For Use With (Application) | Kinase Assay |
Gene Alias | CDK4/Cyclin D1, CND1 |
KinaseFamily | CDK and CHK Kinase Family |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11585026
|
Thermo Scientific™
PV4400 |
10μg |
€ 624.00
10µg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15427182
|
Thermo Scientific™
PR8064A |
100μg |
€ 2905.00
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10482902
|
Thermo Scientific™
PV4436 |
1 mg |
€ 13780.00
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CDK4 functions as a cell-cycle initiator protein. This protein is coexpressed and copurified with CyclinD1.Specifications
NP_000066 I NP_4444284 | |
CDK4/Cyclin D1, CND1 | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
CDK and CHK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Baculovirus | |
Recombinant, Full Length |
For Research Use Only. Not for use in diagnostic procedures.