Learn More
Invitrogen™ CDK4 Monoclonal Antibody (DCS-31.2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA182085
Description
Mouse anti CDK4 antibody clone DCS31.2 recognizes cyclin dependent kinase 4 (Cdk4), a ~34 kDa protein involved in the regulation of cell cycle progression between G1 and S.Mouse anti CDK4 antibody clone DCS31.2 does not cross react with other cdk's and does not co-precipitate the D type cyclins bound to cdk4.
CDK4/cyclin D3 serine/threonine kinase is dimeric protein complex. CDK4 is a member of the cyclin-dependent protein kinase family and is involved in the control of cell proliferation during the G1 phase of cell cycle. CDK4 forms a complex with the D-type cyclins and is inhibited by p16 (cyclin-dependent kinase inhibitor-2). CDK4 can mediate phosphorylation of the C-terminal region of Rb protein leading to an active transcriptional repression of E2F complex. CDC37 and HSP90 can preferentially associate with the fraction of CDK4 not bound to D-type cyclins. SMAD3 is a major physiologic substrate of the G1 cyclin-dependent kinases CDK4 and CDK2. Although CDK6 and CDK4 can both phosphorylate multiple residues in the Rb protein, they do so with different residue selectivity in vitro; CDK6 phosphorylates Thr821 while CDK4 phosphorylates Thr826 on Rb protein.
Specifications
| CDK4 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.08% sodium azide | |
| P11802, P30285, P35426 | |
| Cdk4 | |
| Proprietary Immunogen. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| DCS-31.2 | |
| Unconjugated | |
| Cdk4 | |
| Cdk4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; cyclin-dependent kinase 4; MGC14458; p34>PSK-J3>/cdk4; PSK-J3; serine/threonine kinase; Unknown (protein for MGC:133903) | |
| Mouse | |
| Purified | |
| RUO | |
| 1019, 12567, 94201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.