missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDK4 Recombinant Rabbit Monoclonal Antibody (6N3M6)
GREENER_CHOICE

Product Code. 17175733
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17175733 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17175733 Supplier Invitrogen™ Supplier No. MA535070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CDK4 Recombinant Monoclonal Antibody for ELISA, IHC (P), Western Blot

CDK4/cyclin D3 serine/threonine kinase is dimeric protein complex. CDK4 is a member of the cyclin-dependent protein kinase family and is involved in the control of cell proliferation during the G1 phase of cell cycle. CDK4 forms a complex with the D-type cyclins and is inhibited by p16 (cyclin-dependent kinase inhibitor-2). CDK4 can mediate phosphorylation of the C-terminal region of Rb protein leading to an active transcriptional repression of E2F complex. CDC37 and HSP90 can preferentially associate with the fraction of CDK4 not bound to D-type cyclins. SMAD3 is a major physiologic substrate of the G1 cyclin-dependent kinases CDK4 and CDK2. Although CDK6 and CDK4 can both phosphorylate multiple residues in the Rb protein, they do so with different residue selectivity in vitro; CDK6 phosphorylates Thr821 while CDK4 phosphorylates Thr826 on Rb protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 6N3M6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Cdk4
Gene Accession No. P11802, P30285, P35426
Gene Alias Cdk4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; cyclin-dependent kinase 4; MGC14458; p34>PSK-J3>/cdk4; PSK-J3; serine/threonine kinase; Unknown (protein for MGC:133903)
Gene Symbols Cdk4
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1019, 12567, 94201
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.