missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdk5 Antibody (OTI2H6), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70396MFV450
This item is not returnable.
View return policy
Description
Cdk5 Monoclonal antibody specifically detects Cdk5 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry
Specifications
| Cdk5 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Purified |
| Flow Cytometry | |
| OTI2H6 | |
| Flow Cytometry | |
| Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit | |
| Full length human recombinant protein of human CDK5 (NP_004926) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication, Innate Immunity, Phospho Specific | |
| 1020 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction