missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdk5 Antibody (OTI2H6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70396PCP
This item is not returnable.
View return policy
Description
Cdk5 Monoclonal specifically detects Cdk5 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.
Specifications
| Cdk5 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| CDK5 | |
| Full length recombinant protein of human Cdk5 (NP_004926) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
| Flow Cytometry | |
| OTI2H6 | |
| Flow Cytometry | |
| Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Innate Immunity, Phospho Specific | |
| 1020.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction