missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-767DL594
This item is not returnable.
View return policy
Description
CDK5RAP2 Polyclonal antibody specifically detects CDK5RAP2 in Human samples. It is validated for Western Blot
Specifications
| CDK5RAP2 | |
| Polyclonal | |
| Western Blot | |
| C48, CDK5 activator-binding protein C48, CDK5 regulatory subunit associated protein 2, CDK5 regulatory subunit-associated protein 2, centrosomal protein 215 kDa, Centrosome-associated protein 215, Cep215, DKFZp686B1070, DKFZp686D1070, FLJ10867, KIAA1633microcephaly, primary autosomal recessive 3, MCPH3 | |
| The immunogen recognized by this antibody maps to a region between residues 900 and 950 of human CDK5 Regulatory Subunit Associated Protein 2 using the numbering given in entry NP_060719.4 (GeneID 55755). | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Markers, Chromatin Research | |
| 55755 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction