missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK5RAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555717
This item is not returnable.
View return policy
Description
Immunogen sequence: GDFGVEAVSE GTDSGISAEA AGIDWGIFPE SDSKDPGGDG IDWGDDAVAL QITVLEAGTQ APEGVARGPD ALTLLEYTET RNQFLDELME LEIFLAQRAV ELSEEADVLS VSQFQ Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
This gene encodes a protein that has been reported to function in signaling pathways governing transcriptional regulation and cell cycle progression. It may play a role in tumorigenesis and metastasis. A pseudogene of this gene is located on the long arm of chromosome 20. Alternative splicing results in multiple transcript variants that encode different isoforms. [provided by RefSeq, May 2013].
Specifications
| CDK5RAP3 | |
| Polyclonal | |
| Unconjugated | |
| CDK5RAP3 | |
| 1810007E24Rik; BC002318; C53; C81486; CDK5 activator-binding protein C53; CDK5 activator-binding protein C53; CDK5 regulatory subunit associated protein 3; CDK5 regulatory subunit associated protein IC53-2; CDK5 regulatory subunit-associated protein 3; Cdk5rap3; HSF-27; IC53; IC53-2; ischemic heart CDK5 activator-binding protein C53; LXXLL/leucine-zipper-containing ARFbinding protein; LXXLL/leucine-zipper-containing ARF-binding protein; LZAP; MST016; MSTP016; OK/SW-cl.114; PP1553; Protein HSF-27 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80279 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JB5 | |
| CDK5RAP3 | |
| Recombinant protein corresponding to Human CDK5RAP3. Recombinant protein control fragment (Product #RP-93194). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction