Learn More
Novus Biologicals™ Cdk7 Antibody (MO-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30978-100ug
 View more versions of this product
Product Code. 30066957
  • € 508.00 / 100µg
Description
Cdk7 Monoclonal antibody specifically detects Cdk7 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cdk7
Monoclonal
Unconjugated
Phosphate buffered saline
Mouse
Ammonium sulfate precipitation
RUO
Primary
Human
Purified
Immunohistochemistry, Immunoprecipitation, Western Blot
MO-1
Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation
CAK1Xenopus laevis, cdk-activating kinase), CDK-activating kinase, Cell division protein kinase 7, cyclin-dependent kinase 7, cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, EC 2.7.11.24, p39 Mo15, serine/threonine kinase stk1,39 kDa protein kinase, serine/threonine protein kinase 1, serine/threonine protein kinase MO15, TFIIH basal transcription factor complex kinase subunit
Rabbits were immunized with a recombinant 221 amino acid fragment of the C terminus of Cdk7 recognizing a 40kD protein
100 μg
Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators
1022
Store at -20°C. Avoid freeze/thaw cycles.
IgG2b