Learn More
CDK7 Mouse, anti-Human, CoraLite Plus 488, Clone: 1E7B6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67889-20UL
Description
The CDK7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK7. This antibody recognizes human, mouse antigen. The CDK7 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| CDK7 | |
| 1E7B6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| 39 kDa protein kinase, CAK, CAK1, CDK activating kinase, CDK7, CDKN7, Cell division protein kinase 7, cyclin dependent kinase 7, MO15, p39 Mo15, p39MO15, STK1 | |
| 20 μL | |
| 1022 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |