Learn More
CDK7 Mouse, anti-Human, CoraLite Plus 488, Clone: 1E7B6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67889-20UL
 View more versions of this product
Product Code. 18067085
  • € 145.00 / 20µL
Description
The CDK7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK7. This antibody recognizes human, mouse antigen. The CDK7 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
CDK7
1E7B6
CoraLite Plus 488
Mouse
Primary
Human, Mouse
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
39 kDa protein kinase, CAK, CAK1, CDK activating kinase, CDK7, CDKN7, Cell division protein kinase 7, cyclin dependent kinase 7, MO15, p39 Mo15, p39MO15, STK1
20 μL
1022
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.