missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cdk9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29211
This item is not returnable.
View return policy
Description
Cdk9 Polyclonal antibody specifically detects Cdk9 in Human samples. It is validated for Immunoprecipitation
Specifications
| Cdk9 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| C-2k, CDC2L4CTK1, CDC2-related kinase, Cell division cycle 2-like protein kinase 4, Cell division protein kinase 9, cyclin-dependent kinase 9, cyclin-dependent kinase 9 (CDC2-related kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, PITALRE, serine/threonine protein kinase PITALRE, Serine/threonine-protein kinase PITALRE, TAK | |
| Between 1 and 50 | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Signal Transduction, Virology Bacteria and Parasites | |
| 1025 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction