missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKL1 Antibody (OTI1D9), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71389APC
This item is not returnable.
View return policy
Description
CDKL1 Monoclonal antibody specifically detects CDKL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CDKL1 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1D9 | |
| Western Blot, Immunohistochemistry | |
| CDC2-related kinase 1, cyclin-dependent kinase-like 1, cyclin-dependent kinase-like 1 (CDC2-related kinase), EC 2.7.11, EC 2.7.11.22, KKIALRE, P42, Protein kinase p42 KKIALRE, serine/threonine protein kinase KKIALRE, Serine/threonine-protein kinase KKIALRE | |
| Human recombinant protein fragment corresponding to amino acids 152-357 of human CDKL1(NP_004187) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8814 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction