missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKL1 Monoclonal Antibody (OTI3E8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805786S
This item is not returnable.
View return policy
Description
CDKL1 Monoclonal antibody specifically detects CDKL1 in Human samples. It is validated for Western BlotSpecifications
| CDKL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q00532 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8814 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3E8 | |
| Unconjugated | |
| CDKL1 | |
| 4933411O17Rik; CDC2-related kinase 1; Cdkl1; cyclin dependent kinase like 1; cyclin-dependent kinase-like 1; cyclin-dependent kinase-like 1 (CDC2-related kinase); KKIALRE; P42; protein kinase p42 KKIALRE; serine/threonine protein kinase KKIALRE; serine/threonine-protein kinase KKIALRE; zgc:101002 | |
| Human recombinant protein fragment corresponding to amino acids 152-357 of human CDKL1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction