missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKL1, Mouse anti-Human, Clone: 5B11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008814-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: YPALGLLKGCLHMDPTQRLTCEQLLHHPYFENIREIEDLAKEHNKPTRKTLRKSRKHHCFTETSKLQYLPQLTGSSILPALDNKKYYCDTKKLNYRFPNISpecifications
| CDKL1 | |
| Monoclonal | |
| Unconjugated | |
| cyclin-dependent kinase-like 1 (CDC2-related kinase) | |
| KKIALRE/p42 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 5B11 | |
| In 1x PBS, pH 7.4 | |
| NM_004196 | |
| CDKL1 | |
| CDKL1 (NP_004187, 259 a.a. ∼ 358 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8814 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction