Learn More
CDKL1 Mouse anti-Human, Clone: OTI1H4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF805785
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene product is a member of a large family of CDC2-related serine/threonine protein kinases. It accumulates primarily in the nucleus. Alternative splice variants encoding different protein isoforms have been described, but their full-length nature has not been determined.Specifications
| CDKL1 | |
| Monoclonal | |
| Unconjugated | |
| CDKL1 | |
| KKIALRE, P42 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8814 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q00532 | |
| CDKL1 | |
| Human recombit protein fragment corresponding to amino acids 152-357 of human CDKL1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.