missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDKL2 Monoclonal Antibody (OTI12C2)
GREENER_CHOICE

Product Code. 15996334
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15996334 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15996334 Supplier Invitrogen™ Supplier No. MA526746

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDKL2 Monoclonal Antibody for Western Blot, IHC (P)

The activation of signal transduction pathways by growth factors, hormones and neurotransmitters is mediated by the MAP kinases ERK 1 and ERK 2. ERK proteins are regulated by dual phosphorylation at specific tyrosine and threonine sites mapping within a characteristic Thr-Glu-Tyr motif. The protein kinase p56 KKIAMRE is distantly related to the MAP kinase group of proteins and is closely related to p42 KKIALRE. KKIAMRE is predominantly expressed in testis, kidney, brain and lung. KKIAMRE contains the conserved MAP kinase dual phosphorylation motif in the sequence Thr-Asp-Tyr and is activated by treatment of cells by EGF. However, unlike other MAP kinases, the EGF-stimulated kinase activity does not require phosphorylation of KKIAMRE and KKIALRE in the Thr-Asp-Tyr motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI12C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDKL2
Gene Accession No. Q92772
Gene Alias 5330436L21Rik; AI505225; CDC2-related kinase; Cdkl2; cyclin dependent kinase like 2; cyclin-dependent kinase-like 2; cyclin-dependent kinase-like 2 (CDC2-related kinase); Kkiamre; Kkm; P56; p56 KKIAMRE protein kinase; protein kinase p56 KKIAMRE; serine/threonine protein kinase KKIAMRE; serine/threonine-protein kinase KKIAMRE; testicular tissue protein Li 35
Gene Symbols CDKL2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 291-493 of CDKL2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8999
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.