missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKL5 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32157APC
This item is not returnable.
View return policy
Description
CDKL5 Polyclonal antibody specifically detects CDKL5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| CDKL5 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| cyclin-dependent kinase-like 5, serine/threonine kinase 9, serine/threonine-protein kinase 9 | |
| The immunogen this antibody was made to, maps to a region between residue 350 to 400 of human Cyclin-Dependent Kinase-Like 5 using the numbering given in entry NP_003150.1 | |
| 0.1 mL | |
| Protein Kinase | |
| 6792 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction