Learn More
PARN Mouse anti-Human, Clone: UMAB143, lyophilized, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM800040CF
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a 3′-exoribonuclease, with similarity to the RNase D family of 3′-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| PARN | |
| Monoclonal | |
| Unconjugated | |
| PARN | |
| DAN, DKCB6, PFBMFT4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5073 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Peptide Array, Western Blot | |
| UMAB143 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O95453 | |
| PARN | |
| Human recombit protein fragment corresponding to amino acids 1-266 of human PARN produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.