missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKN1A, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00001026-D02P.100ug
This item is not returnable.
View return policy
Description
Sequence: MSEPAGDVRQNPCGSKACRRLFGPVDSEQLSRDCDALMAGCIQEARERWNFDFVTETPLEGDFAWERVRGLGLPKLYLPTGPRRGRDELGGGRRPGTSPALLQGTAEEDHVDLSLSCTLVPRSGEQAEGSPGGPGDSQGRKRRQTSMTDFYHSKRRLIFSKRKPSpecifications
| CDKN1A | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_000389.1 | |
| CDKN1A | |
| CDKN1A (NP_000380.1, 1 a.a. ∼ 164 a.a) full-length human protein. | |
| RUO | |
| 1026 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| cyclin-dependent kinase inhibitor 1A (p21, Cip1) | |
| CAP20/CDKN1/CIP1/MDA-6/P21/SDI1/WAF1/p21CIP1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction