Learn More
CDKN1C Mouse anti-Human, Clone: 57P06, Abnova™
Mouse monoclonal antibody raised against full length recombinant human CDKN1C.
Brand: Abnova MAB13251.100ug
Description
The protein encoded by this gene is a tight-binding, strong inhibitor of several G1 cyclin/Cdk complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndorome, suggesting that this gene is a tumor suppressor candidate. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]
Specifications
| CDKN1C | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| BWCR/BWS/KIP2/WBS/p57 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 1028 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| 57P06 | |
| Mouse monoclonal antibody raised against full length recombinant human CDKN1C. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| CDKN1C | |
| Recombinant protein corresponding to full length human CDKN1C. | |
| 100 μg | |
| Primary | |
| This monoclonal antibody shows no cross-reaction with CDKN1B. | |
| Store at 4°C. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.