missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKN2A Interacting protein N-Terminal Like Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97396JF549
This item is not returnable.
View return policy
Description
CDKN2A Interacting protein N-Terminal Like Polyclonal specifically detects CDKN2A Interacting protein N-Terminal Like in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CDKN2A Interacting protein N-Terminal Like | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CDKN2A interacting protein N-terminal like, CDKN2A-interacting protein N-terminal-like protein, CDKN2AIP N-terminal-like protein, MGC13017 | |
| Produced in rabbits immunized with E. coli-derived Human CDKN2A Interacting protein N-Terminal Like fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 91368 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction