missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDKN2A (Isoform 3) Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142205
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize the pancreas isoform (NP_478104.1) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This gene generates several transcript variants which differ in their first exons. At least three alternatively spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20 Kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants. This ARF product functions as a stabilizer of the tumor suppressor protein p53 as it can interact with, and sequester, the E3 ubiquitin-protein ligase MDM2, a protein responsible for the degradation of p53. In spite of the structural and functional differences, the CDK inhibitor isoforms and the ARF product encoded by this gene, through the regulatory roles of CDK4 and p53 in cell cycle G1 progression, share a common functionality in cell cycle G1 control. This gene is frequently mutated or deleted in a wide variety of tumors, and is known to be an important tumor suppressor gene. [provided by RefSeq, Sep 2012].
Specifications
| CDKN2A (Isoform 3) | |
| Polyclonal | |
| Unconjugated | |
| CDKN2A | |
| CDKN2A | |
| Peptide with sequence C-ILRKKGRLPGEFSE. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P42771, Q8N726 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1029 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction