missing translation for 'onlineSavingsMsg'
Learn More

CDKN2A Monoclonal Antibody (OTI11D3), TrueMAB™, OriGene

Product Code. 17542504
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17542504 100 μL 100µL
17562504 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17542504 Supplier Origene Technologies Supplier No. TA809881

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

In humans, the multiple tumor suppressor 1 locus (MTS1) encodes two unrelated genes, p16INK4a and p19ARF. Although both act as cell proliferation inhibitors, their mechanisms of action are different. p19ARF works as part of a p53-dependent pathway to counter uncontrolled proliferation and oncogenic signals. Mice lacking the p19ARF gene rapidly develop a broad spectrum of tumors. This result indicates that p19ARF is an important tumor suppressor. In addition, p19ARF helps prevent transforming signals from various onocoproteins via the p53 regulatory loop. These studies and others display the importance of p19ARF in cell cycle control and tumor suppression.

Specifications

Antigen CDKN2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI11D3
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CDKN2A
Gene Accession No. Q8N726
Gene Alias ARF, CDK4I, CDKN2, CMM2, INK4, INK4A, MLM, MTS-1, MTS1, P14, P14ARF, P16, P16-INK4A, P16INK4, P16INK4A, P19, P19ARF, TP16
Gene Symbols CDKN2A
Host Species Mouse
Immunogen Full length human recombit protein of human CDKN2A produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1029
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.