missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM8 Mouse anti-Human, Clone: OTI7A11, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811228S
This item is not returnable.
View return policy
Description
TRPM8 Monoclonal antibody specifically detects TRPM8 in Human samples. It is validated for Western BlotSpecifications
| TRPM8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z2W7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79054 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI7A11 | |
| Unconjugated | |
| TRPM8 | |
| CMR1; Cold menthol receptor 1; cold/menthol receptor 1; long transient receptor potential channel 6; LTRPC6; LTrpC-6; MGC2849; short form of the TRPM8 cationic channel; transient receptor melastatin 8 variant 1; transient receptor melastatin 8 variant 2; transient receptor potential cation channel subfamily M member 8; transient receptor potential cation channel, subfamily M, member 8; transient receptor potential p8; transient receptor potential subfamily M member 8; transient receptor potential-p8; TRP ion channel TRPM8; Trpm8; Trpp8; trp-p8 | |
| Human recombinant protein fragment corresponding to amino acids 1-300 of human TRPM8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction