missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNI Monoclonal Antibody (OTI3F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811170
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin shows the highest similarity with cyclin G. The transcript of this gene was found to be expressed constantly during cell cycle progression. The function of this cyclin has not yet been determined.Specifications
| CCNI | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14094 | |
| CCNI | |
| Human recombit protein fragment corresponding to amino acids 114-377 of human CCNI produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3F10 | |
| Unconjugated | |
| CCNI | |
| CCNI1, CYC1, CYI | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10983 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction