missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRFS1 Mouse anti-Human, Mouse, Rat, Clone: OTI4H8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811461
This item is not returnable.
View return policy
Description
Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.Specifications
| UQCRFS1 | |
| Monoclonal | |
| Unconjugated | |
| UQCRFS1 | |
| RIP1, RIS1, RISP, UQCR5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 291103, 66694, 7386 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P20788, P47985, Q9CR68 | |
| UQCRFS1 | |
| Human recombit protein fragment corresponding to amino acids 79-274 of human UQCRFS1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction