missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKN3 Monoclonal Antibody (OTI1H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503578
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the dual specificity protein phosphatase family. It was identified as a cyclin-dependent kinase inhibitor, and has been shown to interact with, and dephosphorylate CDK2 kinase, thus prevent the activation of CDK2 kinase. This gene was reported to be deleted, mutated, or overexpressed in several kinds of cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| CDKN3 | |
| Monoclonal | |
| 0.64 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16667 | |
| CDKN3 | |
| Full length human recombit protein of human CDKN3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1H5 | |
| Unconjugated | |
| CDKN3 | |
| CDI1, CIP2, KAP, KAP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1033 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction