missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDNF Antibody (104), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90322F
This item is not returnable.
View return policy
Description
CDNF Monoclonal specifically detects CDNF in Human samples. It is validated for ELISA.
Specifications
| CDNF | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 104 | |
| ELISA | |
| arginine-rich, mutated in early stage tumors-like 1, ARMETL1, ARMET-like protein 1, cerebral dopamine neurotrophic factor, Conserved dopamine neurotrophic factor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CDNF (NP_001025125.2; Met1-Leu187). | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 441549 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction