missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553598
This item is not returnable.
View return policy
Description
Immunogen sequence: KSRKSQLSRP EGLNLEPVYF VLSQAGASSL HIQAVTQEHA GKYICEAANE HGTTQAEASL MVVPFETNTK AETVTLPDAA QNDDRSKRDG SETGLLSSFP VKVHPSAVES APEKNASGIS VPDAPIILSP PQTHTPDTYN LVW Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.
CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
Specifications
| CDO | |
| Polyclonal | |
| Unconjugated | |
| CDON | |
| CAM-related/down-regulated by oncogenes; Cd0; Cdo; CDON; Cdon homolog; CDON1; cell adhesion associated, oncogene regulated; cell adhesion molecule-related/down-regulated by oncogenes; HPE11; immunoglobulin superfamily member; immunoglobulin superfamily member CDO; oncogene-regulated cell adhesion molecule; ORCAM; surface glycoprotein, Ig superfamily member | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50937 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4KMG0 | |
| CDON | |
| Recombinant protein corresponding to Human CDO. Recombinant protein control fragment (Product #RP-89967). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction