missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDR2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14563-1-AP-20UL
This item is not returnable.
View return policy
Description
14563-1-AP recognizes a 62 kDa band due to phosphorylation.
Cerebellar degeneration-related protein 2-like (CDR2L), also named as HUMPPA or Paraneoplastic 62 kDa antigen, is a 465 amino acid protein, which belongs to the CDR2 family. CDR2L contains three potential coiled-coil regions. The functions of protein is so far not known. CDR2L is expressed in the brain and localizes in the cytoplasm. The predicted MW of this protein is 53 kDa.Specifications
| CDR2L | |
| Polyclonal | |
| Unconjugated | |
| CDR2L | |
| CDR2L, HUMPPA, Paraneoplastic 62 kDa antigen | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 237988, 30850, 360656 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2A6T1, Q86X02 | |
| Cdr2l | |
| CDR2L Fusion Protein Ag6127 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction