missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDRT15L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568393
This item is not returnable.
View return policy
Description
CDRT15L2 Polyclonal Antibody for IHC (P)
CDRT15L2 is a protein coding gene.
Specifications
| CDRT15L2 | |
| Polyclonal | |
| Unconjugated | |
| CDRT15L2 | |
| CDRT15L2; CMT1A duplicated region transcript 15 protein-like protein; CMT1A duplicated region transcript 15-like 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 256223 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| A8MXV6 | |
| CDRT15L2 | |
| Synthetic peptide corresponding to residues near the N terminal of human CDRT15L2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction