missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDS2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97196JF646
This item is not returnable.
View return policy
Description
CDS2 Polyclonal antibody specifically detects CDS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CDS2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| CDP-DAG synthase 2, CDP-DG synthase 2, CDP-DG synthetase 2, CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2, CDP-diacylglycerol synthase 2, CDP-diglyceride diphosphorylase 2, CDP-diglyceride pyrophosphorylase 2, CDP-diglyceride synthase 2, CDP-diglyceride synthetase 2, CDS 2, CTP:phosphatidate cytidylyltransferase 2, EC 2.7.7, EC 2.7.7.41, FLJ38111, phosphatidate cytidylyltransferase 2 | |
| Produced in rabbits immunized with E. coli-derived Human CDS2 fragment. | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 8760 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction