missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97196-100ul
This item is not returnable.
View return policy
Description
CDS2 Polyclonal antibody specifically detects CDS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CDS2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:10000 | |
| CDP-DAG synthase 2, CDP-DG synthase 2, CDP-DG synthetase 2, CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2, CDP-diacylglycerol synthase 2, CDP-diglyceride diphosphorylase 2, CDP-diglyceride pyrophosphorylase 2, CDP-diglyceride synthase 2, CDP-diglyceride synthetase 2, CDS 2, CTP:phosphatidate cytidylyltransferase 2, EC 2.7.7, EC 2.7.7.41, FLJ38111, phosphatidate cytidylyltransferase 2 | |
| Produced in rabbits immunized with E. coli-derived Human CDS2 fragment. | |
| 100 μg | |
| Endocrinology, Signal Transduction | |
| 8760 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction