missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ V5 Tag Chimeric Recombinant Armenian Hamster Monoclonal Antibody (SV5-P-K)

Armenian Hamster Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551660
This item is not returnable.
View return policy
Description
Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5.
Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).
Specifications
| V5 Tag Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Armenian Hamster | |
| Protein A | |
| RUO | |
| Tag | |
| Antibody | |
| IgG κ |
| ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry | |
| SV5-P-K | |
| Unconjugated | |
| simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag | |
| Pk (V5) Epitope Tag (GKPIPNPLLGLDST). | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction