missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INPP4B Recombinant Rabbit Monoclonal Antibody (23GB2530), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552508
This item is not returnable.
View return policy
Description
Catalyzes the hydrolysis of the 4-position phosphate of phosphatidylinositol 3,4-bisphosphate, inositol 1,3,4- trisphosphate and inositol 1,4-bisphosphate.
Specifications
| INPP4B | |
| Recombinant Monoclonal | |
| 0.115 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15327, Q6P1Y8, Q9QWG5 | |
| INPP4B | |
| A synthesized peptide derived from human INPP4B (1-54AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 23GB2530 | |
| Unconjugated | |
| INPP4B | |
| D-myo-inositol-3,4-bisphosphate 4-phosphohydrolase; E130107I17Rik; inositol polyphosphate 4-phosphatase II; 4-phosphatase II; inositol polyphosphate 4-phosphatase type II; inositol polyphosphate 4-phosphatase type II beta isoform; inositol polyphosphate-4-phosphatase type II 105kD; inositol polyphosphate-4-phosphatase type II B; inositol polyphosphate-4-phosphatase, type II; inositol polyphosphate-4-phosphatase, type II, 105kD; inositol polyphosphate-4-phosphatase, type II, 105kDa; Inpp4b; phosphatidylinositol-3,4-bisphosphate 4-phosphatase; phosphoinositide 4-phosphatase; type II inositol 3,4-bisphosphate 4-phosphatase; type II inositol-3,4-bisphosphate 4-phosphatase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116699, 234515, 8821 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction