missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDw17 Monoclonal Antibody (B392 (HuLy-m13))
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA191271
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Recommended positive controls: Tonsil, Lymph Node, Granulocytes, Platelets.
CDw17, also known as Lactosylceramide (LacCer) or CD17, is a glycosphingolipid present on the surface of various cell types, including leukocytes, erythrocytes, and endothelial cells. It serves as a significant component of the cell membrane, involved in cell-cell interactions, cell adhesion, and signal transduction. Lactosylceramide is synthesized by the enzyme B-1,4-galactosyltransferase, which adds a galactose molecule to glucosylceramide. It acts as a precursor for more complex glycosphingolipids, playing critical roles in numerous physiological processes and immune responses. CDw17 is implicated in several pathological conditions, including atherosclerosis, cancer, and inflammatory diseases. In cancer, CDw17 acts as a mediator for tumor cell adhesion, invasion, and metastasis, making it a potential target for therapeutic intervention. Additionally, due to its role in the immune system, modulation of Lactosylceramide levels can influence immune cell functions and inflammatory responses.
Tekniske data
| CDw17 | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgM |
| Flow Cytometry, Immunocytochemistry | |
| B392 (HuLy-m13) | |
| Unconjugated | |
| alpha 2,3-sialyltransferase V; CD17; CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; Ganglioside GM3 synthase; GM3 synthase; Lactosylceramide; lactosylceramide alpha-2,3-sialyltransferase; SATI; Sialyltransferase 9; sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; GM3 synthase); SIAT9; SIATGM3S; ST3 beta-galactoside alpha-2,3-sialyltransferase 5; ST3 beta-galactoside alpha-23-sialyltransferase 5; ST3Gal V; ST3GAL5; ST3GalV; UNQ2510/PRO5998 | |
| Immunoprecipitated complexes of human detergent PBL lysate and anti-b2m proteins. | |
| 200 μg | |
| Primary | |
| 4°C, do not freeze | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion