missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdx1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98125AF488
This item is not returnable.
View return policy
Description
Cdx1 Polyclonal antibody specifically detects Cdx1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cdx1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915 | |
| Produced in rabbits immunized with E. coli-derived Human Cdx1 fragment. | |
| 0.1 mL | |
| Epigenetics | |
| 1044 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction