missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdx1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49538AF594
This item is not returnable.
View return policy
Description
Cdx1 Polyclonal specifically detects Cdx1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cdx1 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915 | |
| Rabbit | |
| 0.1 mL | |
| 1044 | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CDX1 | |
| A genomic peptide made to an internal region of the human Cdx1 protein (within residues 30-150). [Swiss-Prot P47902] | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction