missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX1 Monoclonal Antibody (OTI3A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810533S
This item is not returnable.
View return policy
Description
CDX1 Monoclonal antibody specifically detects CDX1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CDX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P98177 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4303 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A1 | |
| Unconjugated | |
| FOXO4 | |
| caudal type homeo box 1; caudal type homeo box transcription factor 1; caudal type homeobox 1; caudal type homeobox transcription factor 1; caudal-type homeobox protein 1; caudal-type homeobox protein CDX1; Cdx; CDX1; Cdx-1; Homeobox protein CDX-1 | |
| Human recombinant protein fragment corresponding to amino acids 206-505 of human FOXO4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction