missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdx1 Mouse anti-Human, Clone: CDX1/1650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07425-100ug
This item is not returnable.
View return policy
Description
Cdx1 Monoclonal specifically detects Cdx1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| Cdx1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915 | |
| Recombinant human full-length Cdx1 protein | |
| 100 μg | |
| Epigenetics | |
| 1044 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| CDX1/1650 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction