missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human EGR4 ELISA Kit (Colorimetric)

Product Code. 18603400 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18603400

Brand: Novus Biologicals™ NBP308115

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.09 ng/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Supernatant
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 1961
Gene Symbol EGR4
Quantity 1 Kit
Research Discipline Cell Cycle and Replication
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym AT133, early growth response 4, early growth response protein 4, EGR-4, NGFIC, NGFI-C, PAT133
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.