missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Monoclonal Antibody (OTI5A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802665
This item is not returnable.
View return policy
Description
CDX2 is a caudal type homeobox gene that encodes an intestine-specific transcription factor that is expressed early in intestinal development and may be involved in the regulation of proliferation and differentiation of intestinal epithelial cells. It is expressed in the nuclei of epithelial cells throughout the intestine, from duodenum to rectum. The CDX2 protein is expressed in primary and metastatic colorectal carcinomas and has also been demonstrated in the intestinal metaplasia of the stomach and intestinal-type gastric cancer, while it is not expressed in the normal gastric mucosa. Studies have shown that CDX2 is superior marker compared to CK20.Specifications
| CDX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99626 | |
| CDX2 | |
| Full length human recombit protein of human CDX2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI5A1 | |
| Unconjugated | |
| CDX2 | |
| CDX-3, CDX2/AS, CDX3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1045 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction