missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112380
This item is not returnable.
View return policy
Description
CDX4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a member of a small subfamily of homeobox containing transcription factors. The encoded protein may regulate homeobox gene expression during anteroposterior patterning and hematopoiesis.
Specifications
| CDX4 | |
| Polyclonal | |
| Unconjugated | |
| CDX4 | |
| caudal type homeo box 4; caudal type homeobox 4; caudal type homeobox transcription factor 4; caudal-type homeobox protein 4; Cdx3; Cdx-3; CDX4; Cdx-4; Homeobox protein CDX-4; RGD1561529 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 1046, 12592 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O14627, Q07424 | |
| CDX4 | |
| Recombinant Human Homeobox protein CDX-4 protein (16-174AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction