missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDY1B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16557881
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16557881 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16557881 Supplier Invitrogen™ Supplier No. PA590567

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EKQKKLTWTT TSRIFSNNAR RRTSRSTKAN YSKNSPKTPV TDKHHRSKNR KLFAASKNVR RKAASILSDT KNMEIINSTI ETLAPDSPFD HKTVSGFQKL EKLDPIAADQ QDTVVFKVTE GKLLRDPLSR PGAEQTGIQN KTQIHPLMSQ MSGSVTASMA TGSATRKGIV VLIDPLAANG TTDMHTSVPR VKGGQRNITD DSRDQPFIKK MHFTIRLTES ASTYRDIVVK KEDGFTQIVL STRSTEKNAL NTEVIKEIVN A; Positive Samples: Mouse testis, Rat testis; Cellular Location: Nucleus.

CDY1 containing a chromodomain and a histone acetyltransferase catalytic domain. Chromodomain proteins are components of heterochromatin-like complexes and can act as gene repressors. Histone hyperacetylation is thought to facilitate the transition in which protamines replace histones as the major DNA-packaging protein. This gene encodes a protein containing a chromodomain and a histone acetyltransferase catalytic domain. Chromodomain proteins are components of heterochromatin-like complexes and can act as gene repressors. This protein is localized to the nucleus of late spermatids where histone hyperacetylation takes place. Histone hyperacetylation is thought to facilitate the transition in which protamines replace histones as the major DNA-packaging protein. The human chromosome Y has two identical copies of this gene within a palindromic region; this record represents the more telomeric copy. Chromosome Y also contains a pair of closely related genes in another more telomeric palindrome as well as several related pseudogenes. Two protein isoforms are encoded by transcript variants of this gene. Additional transcript variants have been described, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDY1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias CDY; CDY1; CDY1A; CDY1B; chromodomain protein, Y chromosome, 1; chromodomain protein, Y-linked, 1; testis-specific chromodomain protein on Y; testis-specific chromodomain protein Y 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-320 of human CDY1B (NP_0010038951).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.