missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDY2B Monoclonal Antibody (OTI5E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810457S
This item is not returnable.
View return policy
Description
CDY2B Monoclonal antibody specifically detects CDY2B in Human samples. It is validated for Western BlotSpecifications
| CDY2B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96R06 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10615 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5E1 | |
| Unconjugated | |
| SPAG5 | |
| CDY; CDY2; CDY2A; CDY2B; chromodomain protein, Y chromosome, 2 related; chromodomain protein, Y-linked, 2B; testis-specific chromodomain protein Y 2; testis-specific chromodomain protein Y protein 2 related; Y chromosome chromodomain protein 2B | |
| Human recombinant protein fragment corresponding to amino acids 1-297 of human SPAG5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction