Learn More
Invitrogen™ CDYL2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA550930
Product Code. 15644855
  • € 530.00 / 100µL
Description
CDYL2 Polyclonal Antibody detects endogenous levels of CDYL2 protein.
CDYL2 (Chromodomain Y Like 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includemethylated histone binding. An important paralog of this gene isCDYL.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDYL2
Polyclonal
Unconjugated
CDYL2
1700029M19Rik; 4930453I21Rik; Cdyl2; CDY-like 2; chromodomain protein, Y chromosome-like 2; chromodomain protein, Y-like 2; chromodomain Y-like protein 2; Gm20194; PCCP1; prostate cancer candidate protein 1, PCCP1
Rabbit
Antigen affinity chromatography
RUO
124359
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Q8N8U2
CDYL2
Synthesized peptide derived from the N-terminal region of human CDYL2.