Learn More
Vileda Professional™ CE Duo™ CleanTech™ Sponge
Microfiber cleaning and disinfecting for hard to reach areas. Vileda Professional CE Duo CleanTech Sponge allows you to clean and disinfect hard to reach areas in cleanrooms and controlled environments.
Brand: Vileda Professional™ 152763
Additional Details : Weight : 3.00000kg
Description
- Microfiber overstitched and flame laminated to foam substrate for superior cleaning performance
- Double sided material is RF welded together resulting in clean edges
- Specially formulated Hydrophilic Polyurethane Sponge results in high liquid release for disinfecting
Specifications
Sponge | |
7 in. | |
Autoclavable | |
Yes | |
Cleaning and Disinfecting for Hard to Reach Areas |
Blue, White | |
Non-sterile | |
4 in. | |
6/Pk., 10 Pk./Cs. | |
Polyester Microfiber, Hydrophilic Polyurethane Foam |