Learn More
Vileda Professional™ CE SoftWall Cleaner
Description
- Designed to clean soft wall curtains separating zones within cleanrooms
- Manufactured of electro-polished stainless steel and aluminum
- Completely autoclavable
- Robust contruction and pump action design means higher pressure can be applied during cleaning and easily adjusted as needed
- Unique design allows full contact on curtain from ceiling to floor
Specifications
Specifications
| Product Type | Curtain Cleaner Tool |
| Sterility | Non-sterile |
| Size | 50 in. |
| Quantity | 1 |
| Material | Electropolished Stainless Steel, Aluminum |