missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEA Antibody (PARLAM 4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97718B
This item is not returnable.
View return policy
Description
CEA Monoclonal antibody specifically detects CEA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| CEA | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PARLAM 4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Carcino Embryonic Antigen, Carcinoembryonic antigen | |
| This antibody is a mouse monoclonal IgG1 antibody derived by fusion of Sp 2/0 Ag 14 Mouse myeloma cells with spleen cells from a BABL/c Mouse immunized with isolated Human CEA. The immunogen has been isolated from Human colonic carcinoma cells. | |
| 0.1 mL | |
| Vision | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction