missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM1/CD66a Antibody (B3-17), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36277DL594
This item is not returnable.
View return policy
Description
CEACAM1/CD66a Monoclonal antibody specifically detects CEACAM1/CD66a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
Specifications
| CEACAM1/CD66a | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| B3-17 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
| recombinant human soluble CEACAM-1-Fc (produced in HEK293 cells) | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 634 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction