missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM1/CD66a Antibody (CC1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43390UV
This item is not returnable.
View return policy
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CEACAM1/CD66a | |
| Monoclonal | |
| DyLight 350 | |
| P06731 | |
| CEACAM1 | |
| The immunogen for this antibody was CD66a. | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 634 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| CC1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction